Tlačiť
Desiccator cabinets, vertical, Dry-Keeper™, SP Bel-Art
Dodávateľ: Bel-Art
Dry-Keeper™
468-4121EA 2143.47 EUR
468-4121
Desiccator cabinets, vertical, Dry-Keeper™, SP Bel-Art
Exsikátory Boxy exsikační
Clear and durable PVC desiccator cabinet which has sleek black ABS accents, vertical profile and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials.
  • Three adjustable, removable and perforated shelves allow full circulation of air
  • Shelf rails can be repositioned on any one of 15 steps set at 28 mm intervals
  • Large in-door dial hygrometer for easy visualisation of rH%
This cabinet has a gasketed door which opens to the right and closes with two latches for a tight seal.
Interior opening: 250×430 mm.
Objednávacie informácie: Additional shelves, rails and desiccant to be ordered separately.
Informace o dodávce: Supplied with desiccant tray.