Tlačiť

Anti-HSP90B1 Mouse Monoclonal Antibody (CF555) [clone: 9G10.F8.2]

Dodávateľ: Biotium
BTIUBNC550940-100EA 219.8 EUR
BTIUBNC550940-100 BTIUBNC550940-500
Anti-HSP90B1 Mouse Monoclonal Antibody (CF555) [clone: 9G10.F8.2]
Recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: 9G10.F8.2
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Chicken, Cow, Dog, Guinea Pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR