Tlačiť

Anti-CD7 Mouse Monoclonal Antibody (CF680R) [clone: 124.1D1]

Dodávateľ: Biotium
BNC811046-500EA 596.11 EUR
BNC811046-500 BNC811046-100
Anti-CD7 Mouse Monoclonal Antibody (CF680R) [clone: 124.1D1]
Protilátky
Recognizes a protein of 40 kDa, identified as CD7 (Workshop IV; Code T155). CD7 is a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation.

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 124.1D1
Conjugation: CF680R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR