Tlačiť
Gel permeation chromatography clean-up columns, CLNpak PAE
Dodávateľ: Resonac
SHOWF6714007EA 547.7 EUR
SHOWF6714007 SHOWF6714008 SHOWF7600025
Gel permeation chromatography clean-up columns, CLNpak PAE
Kolony chromatografické
The CLNpak PAE are polyvinyl alcohol based columns suitable for cleaning up high-moisture samples such as blood and bottom sediment. These are highly effective for fractionation of endocrine disruptors in environmental samples.