Tlačiť
Thermostatic mixing valve for body showers
Dodávateľ: Broen Lab
118-0404EA 1183.24 EUR
118-0404
Thermostatic mixing valve for body showers
Pomůcky první pomoci Sprchy
This thermostatic mixing valve is designed for delivering tepid water (20 to 25 °C) for emergency body showers.
  • Fail-safe on hot side: If the cold water is interrupted the built-in anti-scalding device will automatically cease the flow of hot water
  • Fail-safe on cold side: If the hot water is interrupted the built-in intentional leak will allow cold water to flow at a sufficient flow rate to ensure operation
  • Including check valves on both hot and cold supply
Min. working pressure: 1,5 bar
Weight: ~1,5 kg
Please note: Reduced temperature stability at low flow rates.