Tlačiť
UV/Vis spectrophotometer, Libra S50
Dodávateľ: Biochrom
Libra
634-0937EA 0 EUR
634-0937
UV/Vis spectrophotometer, Libra S50
Spektrofotometry a spektrometry Spektrofotometry UV/Vis
Libra S50 is designed to deliver reliable UV/Vis spectrophotometry in environments with multiple users.
  • Colour touch screen user interface with menu driven software and intuitive icons
  • Full PC control with resolution software
  • Built in full suite of software applications with data export methods
  • Convenient USB port enables individual users to load personal methods and transport data
Trace Manager enables the user to manipulate data on the instrument or USB stick. Multi user log allows supervisors to restrict access to software functionality allowing for secure methods.
Life science methods include DNA, RNA and Oligo concentration and purity measurement with wavelength scanning, CyDye™ DNA quantitation, Tm calculation, UV protein concentration measurement, Bradford, Biuret, Lowry or BCA colorimetric methods.