Chcete sa opäť prihlásiť?
Human Acyl CoA binding domain containing protein 6 (ACBD6) ELISA Pair Set
Dodávateľ: Biorbyt
Matched capture and detection antibodies for the preparation of human ACBD6 ELISA assays.
Kit components:
Capture antibody: 0,4 mg/ml of mouse anti-ACBD6 monoclonal antibody. Dilute to a working concentration of 2,0 μg/ml in CBS before coating.
Detection antibody: 0,5 mg/ml mouse anti-ACBD6 monoclonal antibody conjugated to horseradish-peroxidase (HRP). Dilute to working concentration of 0,5 μg/ml in detection antibody dilution buffer before use.
Standard: Each vial contains 150 ng of recombinant ACBD6. Reconstitute with 1 ml detection antibody dilution buffer.
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
Human Acyl CoA binding domain containing protein 6 (ACBD6) ELISA Pair Set, 5 plates 5 BIRBORB145882-5
Human Acyl CoA binding domain containing protein 6 (ACBD6) ELISA Pair Set, 15 plates 15 BIRBORB145882-15
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo