Chcete sa opäť prihlásiť?
Human Acyl CoA binding domain containing protein 6 (ACBD6) ELISA Pair Set
Dodávateľ: Biorbyt
Matched capture and detection antibodies for the preparation of human ACBD6 ELISA assays.
Kit components:
Capture antibody: 0,4 mg/ml of mouse anti-ACBD6 monoclonal antibody. Dilute to a working concentration of 2,0 μg/ml in CBS before coating.
Detection antibody: 0,5 mg/ml mouse anti-ACBD6 monoclonal antibody conjugated to horseradish-peroxidase (HRP). Dilute to working concentration of 0,5 μg/ml in detection antibody dilution buffer before use.
Standard: Each vial contains 150 ng of recombinant ACBD6. Reconstitute with 1 ml detection antibody dilution buffer.
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
Human Acyl CoA binding domain containing protein 6 (ACBD6) ELISA Pair Set, 5 plates 5 BIRBORB145882-5
Human Acyl CoA binding domain containing protein 6 (ACBD6) ELISA Pair Set, 15 plates 15 BIRBORB145882-15
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo