Tlačiť
pH indicator test strips, non bleeding, Dosatest
Dodávateľ: VWR Chemicals
85412.601EA 12.92 EUR
85412.601 85420.601 85416.601 85410.601 85419.601 85421.601 85422.601 85414.601 85423.601 85415.601 85417.601 85418.601 85411.601 85413.601
pH indicator test strips, non bleeding, Dosatest
Papírky testovací Stripy pH indikační
Dosatest test strips provide precise and rapid pH determination, with up to four different colour blocks for each pH value.
  • Easier to work with due to the long handle
  • Colour-fixed reactive pads
  • Brilliant colour chart
pH indicator is chemically bound onto the cellulose fibre preventing bleeding. This effectively protects the sample against contamination and enables measurements to be taken even in weakly buffered or strongly alkaline solutions.
Better access to different types of containers and prevents fingers getting into contact with the sample.
Informace o dodávce: Supplied as a box with 100 test strips 6×85 mm.
Jednoduchý spôsob, ako objaviť chemický produkt
Vyhľadávanie chemikálií podľa metódy (HPLC, GC, Metal traces, XRF atď.), značky (VWRc, Merck atď.) alebo kategórie produktu (normy, chromatografia atď.)
Otestujte ich a presvedčte sa sami