Tlačiť
Labels for TLS 2200®/TLS PC Link™ printers, type B-427
Dodávateľ: Brady
BRDY062259EA 42.59 EUR
BRDY062259 BRDY033891 BRDY062267 BRDY062265 BRDY055153 BRDY018598 BRDY018389 BRDY062261 BRDY018434 BRDY018365 BRDY033879 BRDY018542 BRDY018415 BRDY018535 BRDY018540 BRDY018373
Labels for TLS 2200®/TLS PC Link™ printers, type B-427
Štítky
Labels for the TLS2200® & TLS PC Link™ thermal transfer portable printers.
  • Self-laminating
  • Self-extinguishing
  • Excellent water and oil resistance
  • Excellent abrasion and smudge resistance
  • Matt finish
TLS2200® parts can also be printed on the BMP™71 label printer provided the roll adapter (M71-ADAPT) is used.