Tlačiť
Organic vapour monitors
Dodávateľ: 3M
151-0150EA 81.58 EUR
MMMA3500
Organic vapour monitors
Monitors use the proven scientific principle of diffusion which involves the gradual mixing of molecules of two or more substances.
  • Uses a single charcoal sorbent wafer for collecting organic vapours
  • Accuracy of ±25% at a 95 % confidence level for many workplace contaminants
  • Designed for measuring personal exposure in work areas of industries
18-month shelf life.