Tlačiť
Handheld refractometers, OPTi
Dodávateľ: Bellingham + Stanley
BELL38-48 BELL38-49 BELL38-75
Handheld refractometers, OPTi
Quality instruments for breweries.
  • Shallow prism dish made of 316 stainless steel
  • Zero calibration with distilled water
  • Automated temperature compensation (ATC)
Quality instruments for breweries.
Informace o dodávce: Supplied with two batteries, a multi-language guide, a certificate of calibration and a protective case.