Tlačiť
Melting point apparatus, digital, SMP30
Dodávateľ: Cole Parmer
stuart®
621-2022EA 3564.91 EUR
621-2022
Melting point apparatus, digital, SMP30
Bodotávky
An update of the SMP3, which incorporates some ergonomic features that allow the user to concentrate more comfortably on viewing the sample. The SMP30 allows the user to view the samples and temperature of the block simultaneously without having to refocus the eye. The bright white light LED is a robust source and delivers clean clear illumination of the tubes and sample. An optional printer is available to provide printouts of each result individually including date and time.
  • Accommodates up to three samples simultaneously, designed for easy access to the block for cleaning
  • Temperature display is housed within the eyepiece
  • Viewing head has two way adjustment for comfort
  • Multi-value LCD for important date, time and temperature information
  • BioCote® antimicrobial protection
Informace o dodávce: Supplied with individual calibration certificate and a pack of 100 melting point tubes, open at one end.