Tlačiť
Pressure tanks
Dodávateľ: Sartorius
511-0038EA 0 EUR
511-0038 511-0253 511-0254 511-0252
Pressure tanks
Nádoby reakční Nádoby reakční, cylindrický tvar
These pressure tanks made from 1.4401 (AISI 316) stainless steel serve as reserve containers for pressure filtration, and are also used for the transport, storage and distribution of liquids. As standard, the lid is equipped with a pressure gauge, a safety valve, and a clamp for leak-proof, pressure-resistant closure.
Two handles simplify the handling and the transport. A certificate concerning construction and pressure testing according to the German regulations for pressure tanks is enclosed with every tank (the tanks are specifically designed for pressure, and are not usable as vacuum containers).