Tlačiť
SafetyWasteCaps, fire resistant
Dodávateľ: SCAT Europe
SCTV112912 SCTV112923 SCTV112921
SafetyWasteCaps, fire resistant
Because they are made of pure PTFE and PE-HD, SafetyWasteCaps are resistant against organic solvents, acids and lyes.
  • Resistant against aggressive media
Informace o dodávce: Fittings for capillaries with an outside Ø of ²·³/₃.₂ mm are included.