Tlačiť
SafetyWasteCaps
Dodávateľ: SCAT Europe
590-0762EA 530.51 EUR
SCTV108152 SCTV107260 SCTV107250
SafetyWasteCaps
Available for multiple screw threads and container sizes.
  • Different thread sizes
  • Resistant against aggresive media
  • With connector for exhaust filter
Because they are made of pure PTFE and PE-HD, SafetyWasteCaps are resistant against organic solvents, acids and lyes.