Tlačiť
VWR®, pH Indicator Strips for Coloured Solutions
Dodávateľ: Avantor
35295.607
VWR®, pH Indicator Strips for Coloured Solutions
Papírky testovací Stripy pH indikační
A special pH-test strip that unifies the pH indicator and the colour chart on one strip. Any sample colour has the same effect on both, the reference colours and the reactive pad. This ensures unadulterated and accurate readings in coloured solutions.
  • Ideal pH test for coloured samples
  • Safe and economical, 11×100 mm strips