Tlačiť
Detergents, neodisher® MediClean
Dodávateľ: Dr. Weigert
neodisher®
148-3137EA 116.65 EUR
148-3137 WEIG404313 148-0155 148-0156
Detergents, neodisher® MediClean
Prostředky čistící Detergenty a povrchové čističe
neodisher® MediClean is a detergent with slightly alkaline active cleaning component in a low form formulation.
  • Liquid concentrate for the automated cleaning of thermostable and thermolabile instruments
  • Components: <5% anionic surfactants also enzymes, preservatives (phenoxyethanol, methyl-, ethyl-, butyl-, propyl-, isopropylparaben)
  • Dosage: 0,2 to 1,0%