Tlačiť
UV/Vis spectrophotometer, Jenway® 6305
Dodávateľ: Cole Parmer
Jenway
634-0094EA 7805.18 EUR
634-0094
UV/Vis spectrophotometer, Jenway® 6305
Spektrofotometry a fotometry Spektrofotometry UV/Vis
Compact, high quality, easy to use measurement system for routine analysis in education, environmental analysis, healthcare, food and quality control.
  • Large sample compartment for a wide range of sampling devices to be fitted
  • Simple keypad layout and menu operating protocols
  • Three point wavelength calibration each time the unit is switched on
Informace o dodávce: Supplied with mains lead, 100 disposable cuvettes, 10×10 mm cell holder, interface cable and PC application software on CD-ROM.