Tlačiť
Bioguard
Dodávateľ: Biowest
D1010-1000 D1020-100
Bioguard
Prostředky čistící Dezinfekční prostředek
Bioguard is used to disinfect surfaces of laboratory apparatus and CO₂ incubators. Bioguard prevents contamination with and growth of fungi, bacteria and other sports, mycoplasma, and viruses including HIV and Hepatitis B.
  • Non toxic and Bioguard-S to keep safe all common work surfaces
  • Bioguard-A to disinfect water trays of all common incubators