Tlačiť
Well plates, PP, WebSeal™, barcoded for Vanquish™ UHPLC systems
Dodávateľ: Thermo Fisher Scientific
WebSeal
CRMA60180-P103BEA 62.33 EUR
CRMA60180-P103B CRMA60180-P135B CRMA60180-P211B CRMA60180-P201B CRMA60180-P217B CRMA60180-P206B CRMA60180-P207B CRMA60180-P105B CRMA60180-P216B CRMA60180-P133B
Well plates, PP, WebSeal™, barcoded for Vanquish™ UHPLC systems
Mikrodestičky Mikrodestičky pro HTS
A range of 96-well, PP, chromatography tested, barcoded well plates for the Thermo Scientific™ Vanquish™ and Vanquish™ Flex UHPLC machines. The barcodes indicate the correct orientation of the plate to ensure the correct position in the autosampler and charger unit when loading and running the analytical sequence. Plates cannot be accepted in the wrong orientation.