Tlačiť
Labels for BMP™ 71 printer, type B-425, WorkHorse™
Dodávateľ: Brady
WorkHorse
BRDY114705EA 84.78 EUR
BRDY114705 BRDY114926 BRDY114706
Labels for BMP™ 71 printer, type B-425, WorkHorse™
Štítky
WorkHorse™ white polypropylene labels with acrylic adhesive for applications requiring excellent solvent resistance and print performance.