Tlačiť

Oxid hlinitý, Ecochrom™ Alumina B Activity: I

Dodávateľ: MP Biomedicals

Synonymá: Alumina C

ICNA0202078EA 2868.23 EUR
ICNA0202078
Oxid hlinitý, Ecochrom™ Alumina B Activity: I
Oxid hlinitý

Specific Surface Area: 150 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 10


pH: 10

Vzorec: Al₂O₃
Mw: 101,96 g/mol
Hustota: 3,94 g/cm³ (20 °C) (lit.)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00003424
Číslo CAS: 1344-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR