Tlačiť
Calf native fetuin (from Serum)
Dodávateľ: MP Biomedicals
ICNA0210487401EA 1014.98 EUR
ICNA0210487401 ICNA02104874.1
Calf native fetuin (from Serum)
Proteiny
Preparation method:
A glycoprotein recovered from the globulin fraction of fresh calf serum by ammonium sulphate fractionation.
Fetuin bovine is a protein with the ability to bind to the insulin receptor and inhibit insulin action. Fetuin has been used in studies of type 2 diabetes mellitus (T2DM). In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients. Fetuin has also been used in studies to demonstrate its protective role in the development of arterial calcification. Fetuin also contributes to the attachment and spreading of cells in culture medium.
Fetuin is a protein containing 12 Cys residues, all involved in disulphide bridges. The composition of bovine fetuin (weight %) is polypeptide 74%, hexose 8,3%, hexosamines 5,5%, and sialic acid 8,7%.
Storage temperature.: +4 °C