Tlačiť
Anti-Xylan-1 Mouse Monoclonal Antibody [clone: CCRC-M108]
AGRIAS163135EA 390.82 EUR
AGRIAS163135
Anti-Xylan-1 Mouse Monoclonal Antibody [clone: CCRC-M108]
Type:
Antigen: Xylan 1
Clonality: Monoclonal
Clone: CCRC-M108
Conjugation: MeBSA-conjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Acer pseudoplatanus, Arabidopsis thaliana, Phormium sp., Solanum lycopersicum, Tamarindus indicus