Tlačiť

Anti-SLC22A2 Rabbit Polyclonal Antibody

Dodávateľ: Bioworld Technology
BWRLBS60420EA 316.64 EUR
BWRLBS60420
Anti-SLC22A2 Rabbit Polyclonal Antibody
Protilátky
A synthetic peptide corresponding to residues in Human SLC22A2.

SLC22A2 pAb detects endogenous levels of SLC22A2 protein.

Type: Primary
Antigen: SLC22A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR