Tlačiť

Anti-GSTM2 Rabbit Polyclonal Antibody

Dodávateľ: Bioworld Technology
BWRLBS60429EA 317.33 EUR
BWRLBS60429
Anti-GSTM2 Rabbit Polyclonal Antibody
Protilátky
A synthetic peptide corresponding to residues in Human GSTM2.

GSTM2 pAb detects endogenous levels of GSTM2 protein.

Type: Primary
Antigen: GSTM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR