Tlačiť
Industrial grade cleanroom tapes, EGP™
Dodávateľ: Micronova
MICREGP-125DBEA 1034.51 EUR
MICREGP-125DB
Industrial grade cleanroom tapes, EGP™
Pásky
These industrial grade cleanroom tapes are made of PE with acrylic adhesive.
  • Cleanroom Support Areas
  • Maintenance
  • Clean Build Projects
For applications where ultra-clean tape is not required. The polyethylene backing with acrylic adhesive has low outgassing and good clean-peel properties. Processed in a controlled environment, tapes are wound on a plastic core and sleeve bagged.
Length: 32,9 m
Different colours available.