Every New Brunswick freezer is also equipped with unique S.M.A.R.T. Plus™ diagnostic software to help quickly identify the cause of a fault or set point variance. Both models have four adjustable shelves giving five compartments.
- Easy-to-read front-mounted control panel for temperature setting and alarm display, four PIN security lock
- On/off power switch located behind lockable panel to prevent accidental operation, automatic reset protects microprocessor in event of electrical power spikes
- Designed to fit through standard doorways and lifts
- Remote alarm port for connection to BMS, autodialler and so on
- Audible and visual alarms for high/low temperature, power failure, low battery, filter clean and system failure
Interior construction: High quality 304 L grade stainless steel interior, lockable outer door, stainless steel shelves with five insulated inner doors and heavy-duty, lockable castors. A heated vent prevents vacuum formation.
Range of temperature: −10 to −45 °C, programmable in 1 °C increments, at ambient temperature up to 32 °C
Certifikácie: CE and UL certified. All NBS freezers are CE certified, meet WEEE Directives for disposal, and are 95 to 98% recyclable (by weight).
Objednávacie informácie: Custom built inventory control systems and storage boxes are available; please contact Avantor for details.