Tlačiť
Eyewash station and box
Dodávateľ: Plum
118-1709EA 76.43 EUR
118-1709 118-0082 118-1708
Eyewash station and box
Pomůcky první pomoci Sprchy na výplach očí
Available as either an open station or closed, dust-proof wall box containing 200 ml pH Neutral and 500 ml Plum eyewash. Ideal where there is a risk of eye injuries from acids or alkalis.
  • Open station with mirror and pictogram
  • Wall box with mirror and eyewash sign
  • Wall box ideal for dusty and dirty production areas
Rinsing time: ~2 minutes for pH Neutral, 5 minutes for eyewash
Shelf life: 3 years
W×D×H open station: 230×80×290 mm
W×D×H wall box: 227×110×265 mm