Tlačiť
Eyewash station, pH Neutral
Dodávateľ: Plum
118-0072EA 37.71 EUR
118-0072
Eyewash station, pH Neutral
Pomůcky první pomoci Sprchy na výplach očí
Wall-mounted station containing 1 litre of 4,9% phosphate buffer solution, with pictogram.
  • Suitable for mounting close to any workplace
  • Excellent as a supplement to fixed emergency showers
  • For fast neutralisation of acids and alkali
Rinsing time: 2 minutes
Shelf life: 3 years