Tlačiť
Drum, open-head
Dodávateľ: New Pig
NEWPDRM1147
Drum, open-head
Made of HDPE, blue.
  • Compatible with a wide range of materials, liquids and chemicals
  • Lever lock band can be wire-sealed to prevent theft
These sturdy, lightweight and rustproof HDPE drums are ideal for storage of chemicals, food products and pharmaceuticals.
Certifikácie: Made of USDA and FDA certified and approved resins for direct containment of food products. Also UN rated for shipping solids.