Tlačiť
Descaler for coffee machines, neodisher® CM
Dodávateľ: Dr. Weigert
WEIG326482EA 0 EUR
WEIG326482
Descaler for coffee machines, neodisher® CM
Prostředky čistící Detergenty a povrchové čističe
Reliably removes stubborn lime deposits and dirt particles from all standard coffee and vending machines. Also suitable for the descaling of electric kettles.
Pozor: Not suitable for aluminium, anodised aluminium and light metal alloys.