Tlačiť
Descaler for surfaces and appliances, caraform® special
Dodávateľ: Dr. Weigert
WEIG322413EA 0 EUR
WEIG322413 WEIG322449 WEIG322442 148-0149
Descaler for surfaces and appliances, caraform® special
Prostředky čistící Detergenty a povrchové čističe
Reliably removes lime and other acid-soluble soiling as well as enclosed dirt particles. Universally applicable for kettles bains-maries, sinks, cooking equipment and surfaces.
  • Liquid concentrate
  • With the EU Ecolabel excellent acid cleaner
  • Based on citric acid
Suitable for all materials such as glass, porcelain, plastic and stainless steel.
Pozor: Surfaces that come into contact with food must be rinsed with drinking water after each cleaning and disinfection to remove residues.