Tlačiť
Solubility index mixer
Dodávateľ: Funke-Gerber
GERB7610EA 2840.94 EUR
GERB7610
Solubility index mixer
Homogenizátory Míchačky laboratorní pro standardní analýzu
Solubility index mixer is used for determining the solubility of milk, cream and whey powder.
Certifikácie: In accordance with ADPI and DLG regulations.
Objednávacie informácie: Replacement parts can be ordered separately.
Informace o dodávce: Mixer is supplied with special motor, glass mixing bowl, stainless steel stirrer, timer and continuous operation switch.