Tlačiť
Protein purification kits, Vivapure® Mini/Maxiprep
Dodávateľ: Sartorius
SARTVS-ARGMAXIK SARTVS-MCMINI24 SARTVS-ARGBUFPK SARTVS-ARGMINIK SARTVS-ARSTPKA2 SARTVS-ARSTPKG2 SARTVS-ARAMINIB SARTVS-MCMAXIK SARTVS-ARAMINIK SARTVS-MCBUFPK SARTVS-ARGMAXIB SARTVS-MCMAXIB SARTVS-ARAMAXIB SARTVS-MCST04 SARTVS-ARABUFPK
Protein purification kits, Vivapure® Mini/Maxiprep
Vivapure® Miniprep and Maxiprep kits are spin column based kits for fast and effective purification of His-tagged proteins and antibodies.
  • Ready to use
  • Spin columns have the advantage of speed over gravity drip columns and batch protocols
Objednávacie informácie: Available as kits, individual devices and bulk packs.