Tlačiť
Detergent for manual cleaning, liquid, neodisher® LM 2
Dodávateľ: Dr. Weigert
148-3182EA 208.77 EUR
WEIG440330
Detergent for manual cleaning, liquid, neodisher® LM 2
  • Mildly alkaline liquid concentrate for washing surgical instruments and laboratory glassware by hand or in ultrasonic and immersion baths
  • Components: < 5% amphoteric surfactants, anionic surfactants, 15 - 30% phosphates
  • Dosage: 0,5 - 3,0%