Tlačiť
Labels, tags, energy source, gas
Dodávateľ: Brady
BRDY138419EA 83.99 EUR
BRDY138419 BRDY138759 BRDY138418 BRDY138758 BRDY138757 BRDY138422 BRDY138421 BRDY138420 BRDY138760 BRDY138756
Labels, tags, energy source, gas
Štítky
These gas energy source tags are constructed of PP with round corners with one hole.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Purple on white
Certifikácie: ATEX
Balenie: 25 tags per pack.