Tlačiť
Labels, tags, energy Source, glycol
Dodávateľ: Brady
BRDY138804EA 83.99 EUR
BRDY138804 BRDY138803 BRDY138802 BRDY138808 BRDY138807 BRDY138806 BRDY138805 BRDY138444 BRDY138446 BRDY138445
Labels, tags, energy Source, glycol
Štítky
These glycol energy source tags are constructed of PP with round corners with one hole.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Green on white
Certifikácie: ATEX
Balenie: 25 tags per pack.