Tlačiť
Labels, tags, Energy Source, hermal
Dodávateľ: Brady
BRDY138438EA 84.17 EUR
BRDY138438 BRDY138781 BRDY138780 BRDY138785 BRDY138784 BRDY138783 BRDY138782 BRDY138437 BRDY138434 BRDY138786
Labels, tags, Energy Source, hermal
Štítky
These thermal energy source tags are constructed of PP with round corners with one hole.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Green on white
Certifikácie: ATEX
Balenie: 25 tags per pack.