Tlačiť
Wall switch lockout, Prinzing
Dodávateľ: Brady
BRDY830892EA 19.56 EUR
BRDY830892
Wall switch lockout, Prinzing
This clear plastic lockout uses existing wall plate screws to mount.
  • Made of strong, clear PC
  • Patented non-hinge design
Objednávacie informácie: Lock is not included.