Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EHD2 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 60 kDa band observed in human heart lysates (calculated MW of 61.2 kDa according to NP_055416.2). In transfected HEK293 transiently expressing EHD2 a band of approximately 62 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.003-0.01 μg/mL.

EHD2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EHD2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EHD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-523
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol EHD2
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
Concentration 500 ug/mL
Immunogen EHD2 antibody was raised against a 15 amino acid synthetic peptide near the C-Terminus of EHD2.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo
 

Nový vzhľad a obnovený záväzok

Postavené na funkciách a zdrojoch, ktoré poznáte, optimalizované s inováciami od spoločnosti Avantor®. Naša nová značka symbolizuje agilitu a našu schopnosť prispôsobiť sa potrebám zákazníkov.

ĎALŠIE INFORMÁCIE ZAČAŤ NAKUPOVAŤ