Tlačiť

Anti-EIF2C1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-525EA 616.08 EUR
PRSI45-525
Anti-EIF2C1 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:16000. Western Blot: In transfected HEK293 transiently expressing human EIF2C1bands of approximately 110 kDa and 35 kDa are observed. The 110 kDa band is not observed in the non-transfected HEK293. The calculated molecular size is 97.2 kDa according to NP_036331.1. Recommended concentration, 0.3-1 μg/mL.

EIF2C1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EIF2C1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EIF2C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR