Tlačiť

Anti-ELF1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-526EA 616.08 EUR
PRSI45-526
Anti-ELF1 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 70 kDa band observed in human tonsil, lymph node and in Jurkat T-cell line lysates (calculated MW of 67.5 kDa according to NP_758961.1). Recommended concentration: 0.3-1.0 μg/mL.

ELF1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ELF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ELF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR