Tlačiť

Anti-ERC1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-529EA 614.75 EUR
PRSI45-529
Anti-ERC1 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 85 kDa band observed in human pancreas and testis lysates (calculated MW of 82 kDa according to NP_829882, isoform gamma). A second band of 37 kDa is detected. Both bands are successfully blocked by incubation with the immunizing peptide. No proteins of approximatelyimately 37 kDa were found after a BLASTP search in NCBI with the immunizing peptide as a probe. Recommended concentration: 0.1-0.3 μg/mL.

ELKS antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ELKS antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ERC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR