Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FBXO43 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 55 kDa band observed in human kidney fibroblast cell line HEK293 lysates (calculated MW of 51.4 kDa according to NP_001025031.2). Recommended concentration: 0.1-0.3 μg/mL.

EMI2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EMI2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: FBXO43
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Rat

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-533
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol FBXO43
Antigen name F-box Protein 43
Reactivity Rat
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms ERP1, FBX43, EMI2
Concentration 500 ug/mL
Immunogen EMI2 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of EMI2.
Form Liquid
Tested applications Fluorescence-Activated Cell Sorting
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo