Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-TXNDC5 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 30 kDa band observed in mouse and rat brain lysates (calculated MW of 34.2 kDa according to human NP_001418.2 and of 33.8 kDa according to mouse NP_034264.1 and of 33.7 kDa according to rat NP_001102684.1). Recommended concentration: 0.1-0.3 μg/mL.

EndoPDI antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EndoPDI antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: TXNDC5
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Mouse, Rat

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-536
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol TXNDC5
Antigen name Thioredoxin Domain Containing 5
Reactivity Rat, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
Antigen synonyms PDIA15, UNQ364, HCC2, ENDOPDI, STRF8, ERP46, HCC-2
Concentration 500 ug/mL
Immunogen EndoPDI antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of EndoPDI.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo