Tlačiť

Anti-ENPP1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-538EA 616.08 EUR
PRSI45-538
Anti-ENPP1 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:32000. Western Blot: In transfected HEK293 transiently expressing ENPP1 a band of approximately 100 kDa is observed. This band is not observed in the non-transfected HEK293. The calculated molecular size is 105 kDa according to NP_006199.2. Recommended concentration, 1-3 μg/mL. Immunohistochemistry: In paraffin embedded liver shows membranous staining in many hepatocytes. This antibody has also been successfully used on human pancreas showing membranous staining of acinar cells. Recommended concentration, 1-2 μg/mL.

ENPP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ENPP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ENPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR