Tlačiť

Anti-PTGER1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-539EA 616.08 EUR
PRSI45-539
Anti-PTGER1 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:128,000.

EP1 Receptor antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EP1 Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTGER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR