Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EPHB2 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:4,000. Approximately 60 kDa band observed in human lung and placenta lysates (calculated MW of 110 kDa according to NP_059145.2 and NP_004433.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Litterst et al, J Biol Chem. 2007 Apr 10; PMID: 17428795). Recommended concentration: 1-3 μg/mL.

EPHB2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EpH B2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: EPHB2
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Human

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-544
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol EPHB2
Antigen name EPH Receptor B2
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
Antigen synonyms DRT, PCBC, EK5, CAPB, Hek5, Tyro5, EPHT3, ERK
Concentration 500 ug/mL
Immunogen EPHB2 antibody was raised against a 12 amino acid synthetic peptide near the internal region of EPHB2.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo