Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EPM2AIP1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 22-23 kDa band in human brain lysates after 1 μg/mL antibody staining.4 kDa according to NP_055620. The 22-23 kDa band was successfully blocked by incubation with the immunizing peptide.

EPM2A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EPM2A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: EPM2AIP1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Dog, Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-547
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol EPM2AIP1
Reactivity Human, Mouse, Dog, Rat
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
Concentration 500 ug/mL
Immunogen EPM2A antibody was raised against a synthetic peptide of EPM2A.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo